Merck
Probiotic consumption decreases the number of osteoclasts during orthodontic movement in mice.
Archives of oral biology (2017-03-11)
Camila Alessandra Pazzini, Luciano José Pereira, Tarcília Aparecida da Silva, Carina Cristina Montalvany-Antonucci, Soraia Macari, Leandro Silva Marques, Saul Martins de Paiva
ABSTRAKT
The aim of the present study was to investigate the effect of probiotic (Bacillus Subtilis) supplementation on bone remodelling induced by mechanical loading. C57BL/6 mice were divided in two groups: (1) Probiotic and (2) Vehicle (water). The probiotic (1.5×10 OTM was similar between groups (with and without probiotic supplementation) (p=0.46). The number of TRAP-positive cells increased (p<0.01) on the experimental side (where the spring coil was installed) in comparison to the control side in both groups. However, the number of osteoclasts decreased (p˂0.01) in the probiotic group, in comparison to the vehicle group. There was an increase in the number of osteoblasts (p˂0.05) in both the Vehicle and Probiotic groups on the side under OTM, independent of probiotic supplementation. Oral Supplementation with a probiotic influenced the number of osteoclasts adjacent to the tooth root during orthodontic movement in mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)