Merck
Evaluation of Legionella real-time PCR against traditional culture for routine and public health testing of water samples.
Journal of applied microbiology (2017-04-05)
S Collins, D Stevenson, J Walker, A Bennett
ABSTRAKT
To evaluate the usefulness of Legionella qPCR alongside traditional culture for enumeration of Legionella from water samples as part of both routine and public health investigation testing. Routine water samples (n = 2002) and samples from public health investigations (n = 215) were analysed by culture and qPCR for Legionella spp., Legionella pneumophila and L. pneumophila sg-1. A negative qPCR result was highly predictive of a negative culture result for all water systems (negative predictive values, NPV from 97·4 to 100%). Positive predictive values (PPV) were lower (0-50%). Results for qPCR were generally larger than culture with average log The high NPV of qPCR supports its use to rapidly screen out negative samples without culture. Alert and action levels for Legionella qPCR for HCWS are proposed. Quantitative PCR will be a valuable tool for both routine and public health testing. This study generated comparative data of >2000 water samples by qPCR and culture. Action and alert levels have been recommended that could enable duty holders to interpret qPCR results to facilitate timely Legionella control and public health protection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Cysteine, technical grade