Merck
Cold tolerance of the Asian tiger mosquito Aedes albopictus and its response to epigenetic alterations.
Journal of insect physiology (2017-04-12)
Aljoscha Kreß, Ann-Marie Oppold, Ulrich Kuch, Jörg Oehlmann, Ruth Müller
ABSTRAKT
Phenotypic plasticity is considered as one of the key traits responsible for the establishment of populations of the invasive mosquito Aedes albopictus, an important vector of viral and parasitic pathogens. The successful spread of this species to higher altitudes and latitudes may be explained by its ability to rapidly induce a heritable low-temperature phenotype (cold hardiness in eggs). As a result of the low genetic diversity of founder populations, an epigenetic short-term mechanism has been suggested as the driver of this diversification. We investigated if random epigenetic alterations promoted the cold hardiness of Ae. albopictus eggs from a transgenerational study of two epigenetic agents (genistein and vinclozolin). To this end, we evaluated changes in lethal time for 50% of pharate larvae (Lt
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Vinclozolin, PESTANAL®, analytical standard