Merck
  • Carotid artery stenosis with a high-intensity signal plaque on time-of-flight magnetic resonance angiography and association with evidence of intraplaque hypoxia.
Carotid artery stenosis with a high-intensity signal plaque on time-of-flight magnetic resonance angiography and association with evidence of intraplaque hypoxia.
Journal of neurosurgery (2016-07-02)
Atsushi Ogata, Masatou Kawashima, Tomihiro Wakamiya, Masashi Nishihara, Jun Masuoka, Yukiko Nakahara, Ryo Ebashi, Kohei Inoue, Yukinori Takase, Hiroyuki Irie, Tatsuya Abe
ABSTRAKT
OBJECTIVE Hypoxia induces angiogenesis and plays a major role in the progression of carotid plaques. During carotid intervention, plaques with high-intensity signals on time-of-flight (TOF) magnetic resonance angiography (MRA) often cause ischemic stroke and embolic complications. However, the role of intraplaque hypoxia before carotid endarterectomy (CEA) and carotid artery stenting is not presently understood. In this study the authors aimed to investigate the relationship between intraplaque hypoxia and MRA findings. METHODS Nineteen consecutive patients with 20 carotid artery stenoses who underwent CEA at Saga University Hospital between August 2008 and December 2014 were enrolled in the study. The expressions of hypoxia-inducible transcription factor-1α (HIF-1α) and vascular endothelial growth factor (VEGF) were analyzed by immunohistochemical analysis. In addition, the relationship between the findings on TOF MRA and pathology for the carotid plaques was analyzed. RESULTS High-intensity plaques on TOF MRA showed higher expression levels of HIF-1α (p = 0.015) and VEGF (p = 0.007) compared with isointensity plaques. The rate of intraplaque hemorrhage (IPH) on TOF MRA was also significantly higher in the high-intensity plaques than in the isointensity plaques (p = 0.024). Finally, the mean number of neovessels was significantly higher in those without plaque hemorrhage than in those with plaque hemorrhage (p = 0.010). CONCLUSIONS Plaques with high-intensity signals on TOF MRA were associated with IPH and evidence of intraplaque hypoxia. This fact may represent an opportunity to establish novel therapeutic agents targeting intraplaque hypoxia.