Merck
Synthesis of n-type Mg
Nanoscale research letters (2017-05-13)
Keiko Kikuchi, Kodai Yamamoto, Naoyuki Nomura, Akira Kawasaki
ABSTRAKT
Magnesium silicide (Mg
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D.E.R. 332, used as embedding medium