Merck
ELF18-INDUCED LONG-NONCODING RNA Associates with Mediator to Enhance Expression of Innate Immune Response Genes in Arabidopsis.
The Plant cell (2017-04-13)
Jun Sung Seo, Hai-Xi Sun, Bong Soo Park, Chung-Hao Huang, Shyi-Dong Yeh, Choonkyun Jung, Nam-Hai Chua
ABSTRAKT
The plant immune response is a complex process involving transcriptional and posttranscriptional regulation of gene expression. Responses to plant immunity are initiated upon the perception of pathogen-associated molecular patterns, including peptide fragment of bacterial flagellin (flg22) or translation elongation factor Tu (elf18). Here, we identify an
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%