Merck
  • Scavenging free radicals and soaring osteoinduction by extra cellular matrix protein-based nanocomposites on degenerative bone treatments.
Scavenging free radicals and soaring osteoinduction by extra cellular matrix protein-based nanocomposites on degenerative bone treatments.
Materials science & engineering. C, Materials for biological applications (2017-05-24)
Kavitha Kandiah, Navaneethan Duraisamy, Vinoth Amirthalingam, Balagurunathan Ramasamy
ABSTRAKT
A number of materials are now available to alleviate the ever-growing bone disruption. However, these are inadequate and inappropriate for addressing issues associated natural process of aging and degeneration of bone due to diseases. This study advances the existing material and offers more privileged and synergistically active remedy for these conditions. Here, they are three different nano-composites prepared such as nano-TiO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)