Merck
Papain-functionalized gold nanoparticles as heterogeneous biocatalyst for bioanalysis and biopharmaceuticals analysis.
Analytica chimica acta (2017-03-25)
Siyao Liu, Markus Höldrich, Adrian Sievers-Engler, Jeannie Horak, Michael Lämmerhofer
ABSTRAKT
Surface-modified gold nanoparticles (GNPs) were synthesized via layer-by-layer process with alternating cationic polyallylamine and anionic poly(acrylic acid) polyelectrolyte layers leading to a highly hydrophilic biocompatible shell supporting colloidal stability. Afterwards, papain was covalently immobilized on the modified GNPs via amide coupling between the amino groups on papain and the terminal carboxylic groups of the modified GNPs by using N-(3-dimethylaminopropyl)-N'-ethylcarbodiimide and N-hydroxysulfosuccinimide sodium as coupling agents. The resultant papain-functionalized gold nanoparticles were characterized by surface plasmon resonance, dynamic light scattering and zeta potential measurements. The new technology resonant mass measurement was applied for determining the average number of papain molecules immobilized per GNP by measurement of the single nanoparticle buoyant mass in the range of femtograms. The activity of the immobilized enzyme was estimated by determination of the kinetic parameters (K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Hydroxymethyl-1,3-propanediol, 97%
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)