- Annexin A8 promotes VEGF-A driven endothelial cell sprouting.
Annexin A8 promotes VEGF-A driven endothelial cell sprouting.
Cell adhesion & migration (2017-01-07)
Nicole Heitzig, Benjamin F Brinkmann, Sophia N Koerdt, Gonzalo Rosso, Victor Shahin, Ursula Rescher
PMID28060564
ABSTRAKT
The physiological and pathological process of angiogenesis relies on orchestrated endothelial cell (EC) adhesion, migration and formation of new vessels. Here we report that human umbilical vein endothelial cells (HUVECs) deficient in Annexin A8 (AnxA8), a member of the annexin family of Ca
MATERIAŁY