Merck
Maternal exposure to perfluorooctanoic acid inhibits luteal function via oxidative stress and apoptosis in pregnant mice.
Reproductive toxicology (Elmsford, N.Y.) (2017-02-22)
Yilu Chen, Ling Zhou, Jingjie Xu, Lu Zhang, Mo Li, Xingxing Xie, Yajuan Xie, Dan Luo, Dalei Zhang, Xiaochun Yu, Bei Yang, Haibin Kuang
ABSTRAKT
Perfluorooctanoic acid (PFOA) is a synthetic perfluorinated compound, which has been reported to exert adverse effect on the pregnancy. However, whether it is associated with alteration of luteal function remains unknown. Mice were administered PFOA by gavage from gestational days (GD) 1-7 or 13. PFOA treatment did not significantly affect numbers of embryo implantation. Nevertheless, on GD 13, 10mg/kg PFOA treatment significantly increased numbers of resorbed embryo. Furthermore, PFOA exposure markedly reduced serum progesterone levels but did not affect estradiol levels. Treatment also showed concomitant decreases in transcript levels for key steroidogenic enzymes, and reduced numbers and sizes of corpora lutea. In addition, PFOA administration inhibited activities of superoxide dismutase and catalase, and increased generation of hydrogen peroxide and malondialdehyde, and down-regulated level of Bcl-2 and up-regulated p53 and BAX proteins. In conclusion, PFOA exposure significantly inhibits luteal function via oxidative stress and apoptosis in pregnant mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Kwas perfluorooktanowy, 95%