Merck
Tetra-2,3-pyrazinoporphyrazines with externally appended pyridine rings. 17. Photosensitizing properties and cellular effects of Zn
Journal of photochemistry and photobiology. B, Biology (2017-03-18)
Elisa Viola, Maria Pia Donzello, Fabiola Sciscione, Kous Shah, Claudio Ercolani, Giuseppe Trigiante
ABSTRAKT
The photosensitizing activity of two multiply charged porphyrazine derivatives, i.e. the Zn
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
29H,31H-Phthalocyanine, β-form, 98%