Merck
Strong enhancement of electrical conductivity in two-dimensional micrometer-sized RuO
Nanoscale (2017-05-18)
Somi Yoo, Jeongmin Kim, Hongjae Moon, Se Yun Kim, Dong-Su Ko, Weon Ho Shin, Sungwoo Hwang, Doh Won Jung, Soohwan Sul, Chan Kwak, Jong Wook Roh, Wooyoung Lee
ABSTRAKT
The enhancement in electrical transport properties of exfoliated individual RuO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ruthenium(IV) oxide, 99.9% trace metals basis