Merck
Gli1-induced deubiquitinase USP48 aids glioblastoma tumorigenesis by stabilizing Gli1.
EMBO reports (2017-06-18)
Aidong Zhou, Kangyu Lin, Sicong Zhang, Li Ma, Jianfei Xue, Saint-Aaron Morris, Kenneth D Aldape, Suyun Huang
ABSTRAKT
Aberrant activation of the Hedgehog (Hh) signaling pathway drives the tumorigenesis of multiple cancers. In this study, we screened a panel of deubiquitinases that may regulate the Hh pathway. We find that deubiquitinase USP48 activates Gli-dependent transcription by stabilizing Gli1 protein. Mechanistically, USP48 interacts with Gli1 and cleaves its ubiquitin off directly. In glioblastoma cells, knockdown of USP48 inhibits cell proliferation and the expression of Gli1's downstream targets, which leads to repressed glioblastoma tumorigenesis. Importantly, USP48's effect on cell proliferation and tumorigenesis depends to some extent on Gli1. In addition, we find that the Sonic Hedgehog (SHH) pathway induces USP48 expression through Gli1-mediated transcriptional activation, which forms thus a positive feedback loop to regulate Hh signaling. In human glioblastoma specimens, the expression levels of USP48 and Gli1 proteins are clinically relevant, and high expression of USP48 correlates with glioma malignancy. In summary, our study reveals that the USP48-Gli1 regulatory axis is critical for glioma cell proliferation and glioblastoma tumorigenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
MISSION® esiRNA, targeting human USP48
Sigma-Aldrich
Przeciwciało anty-HA, Upstate®, from mouse