- Caffeine inhibits hypothalamic A
Caffeine inhibits hypothalamic A
Nature communications (2017-06-28)
Liufeng Wu, Jia Meng, Qing Shen, Yi Zhang, Susu Pan, Zhuo Chen, Ling-Qiang Zhu, Youming Lu, Yuan Huang, Guo Zhang
PMID28654087
ABSTRAKT
Caffeine, an antagonist of the adenosine receptor A