Merck
PGBD5 promotes site-specific oncogenic mutations in human tumors.
Nature genetics (2017-05-16)
Anton G Henssen, Richard Koche, Jiali Zhuang, Eileen Jiang, Casie Reed, Amy Eisenberg, Eric Still, Ian C MacArthur, Elias Rodríguez-Fos, Santiago Gonzalez, Montserrat Puiggròs, Andrew N Blackford, Christopher E Mason, Elisa de Stanchina, Mithat Gönen, Anne-Katrin Emde, Minita Shah, Kanika Arora, Catherine Reeves, Nicholas D Socci, Elizabeth Perlman, Cristina R Antonescu, Charles W M Roberts, Hanno Steen, Elizabeth Mullen, Stephen P Jackson, David Torrents, Zhiping Weng, Scott A Armstrong, Alex Kentsis
ABSTRAKT
Genomic rearrangements are a hallmark of human cancers. Here, we identify the piggyBac transposable element derived 5 (PGBD5) gene as encoding an active DNA transposase expressed in the majority of childhood solid tumors, including lethal rhabdoid tumors. Using assembly-based whole-genome DNA sequencing, we found previously undefined genomic rearrangements in human rhabdoid tumors. These rearrangements involved PGBD5-specific signal (PSS) sequences at their breakpoints and recurrently inactivated tumor-suppressor genes. PGBD5 was physically associated with genomic PSS sequences that were also sufficient to mediate PGBD5-induced DNA rearrangements in rhabdoid tumor cells. Ectopic expression of PGBD5 in primary immortalized human cells was sufficient to promote cell transformation in vivo. This activity required specific catalytic residues in the PGBD5 transposase domain as well as end-joining DNA repair and induced structural rearrangements with PSS breakpoints. These results define PGBD5 as an oncogenic mutator and provide a plausible mechanism for site-specific DNA rearrangements in childhood and adult solid tumors.