Merck
RNA target profiles direct the discovery of virulence functions for the cold-shock proteins CspC and CspE.
Proceedings of the National Academy of Sciences of the United States of America (2017-06-15)
Charlotte Michaux, Erik Holmqvist, Erin Vasicek, Malvika Sharan, Lars Barquist, Alexander J Westermann, John S Gunn, Jörg Vogel
ABSTRAKT
The functions of many bacterial RNA-binding proteins remain obscure because of a lack of knowledge of their cellular ligands. Although well-studied cold-shock protein A (CspA) family members are induced and function at low temperature, others are highly expressed in infection-relevant conditions. Here, we have profiled transcripts bound in vivo by the CspA family members of
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent