Merck
Simultaneous lipolysis/permeation in vitro model, for the estimation of bioavailability of lipid based drug delivery systems.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2017-05-10)
Hanady Ajine Bibi, René Holm, Annette Bauer-Brandl
ABSTRAKT
The simultaneous processes of lipid digestion and absorption together determine the oral bioavailability of drugs incorporated into lipid based drug delivery systems (LBDDS). A number of slightly different protocols for in vitro lipolysis are widely accepted; however, the permeation process has so far not been included into the models due to the harsh conditions of lipid digestion compromising permeation barriers. The present study for the first time combines biomimetic permeation and lipolysis of LBDDS. The focus of the current work was on the functional stability of the barrier - Permeapad
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
4-Bromophenylboronic acid, ≥95.0%