Merck
Effect of low-intensity pulsed ultrasound on osteogenic human mesenchymal stem cells commitment in a new bone scaffold.
Journal of applied biomaterials & functional materials (2017-05-10)
Valeria Carina, Viviana Costa, Lavinia Raimondi, Stefania Pagani, Maria Sartori, Elisa Figallo, Stefania Setti, Riccardo Alessandro, Milena Fini, Gianluca Giavaresi
ABSTRAKT
Bone tissue engineering is helpful in finding alternatives to overcome surgery limitations. Bone growth and repair are under the control of biochemical and mechanical signals; therefore, in recent years several approaches to improve bone regeneration have been evaluated. Osteo-inductive biomaterials, stem cells, specific growth factors and biophysical stimuli are among those. The aim of the present study was to evaluate if low-intensity pulsed ultrasound stimulation (LIPUS) treatment would improve the colonization of an MgHA/Coll hybrid composite scaffold by human mesenchymal stem cells (hMSCs) and their osteogenic differentiation. LIPUS stimulation was applied to hMSCs cultured on MgHA/Coll hybrid composite scaffold in osteogenic medium, mimicking the microenvironment of a bone fracture. hMSCs were seeded on MgHA/Coll hybrid composite scaffold in an osteo-inductive medium and exposed to LIPUS treatment for 20 min/day for different experimental times (7 days, 14 days). The investigation was focused on (i) the improvement of hMSCs to colonize the MgHA/Coll hybrid composite scaffold by LIPUS, in terms of cell viability and ultrastructural analysis; (ii) the activation of MAPK/ERK, osteogenic (ALPL,COL1A1,BGLAP,SPP1) and angiogenetic (VEGF, IL8) pathways, through gene expression and protein release analysis, after LIPUS stimuli. LIPUS exposure improved MgHA/Coll hybrid composite scaffold colonization and induced in vitro osteogenic differentiation of hMSCs seeded on the scaffold. This work shows that the combined use of new biomimetic osteo-inductive composite and LIPUS treatment could be a useful therapeutic approach in order to accelerate bone regeneration pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Human KIT / Mast/Stem Cell Growth Factor Receptor Kit ELISA Kit