Merck
The Probiotic VSL#3 Modulates Colonic Macrophages, Inflammation, and Microflora in Acute Trinitrobenzene Sulfonic Acid Colitis.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2017-07-12)
Raymond A Isidro, Abdon Lopez, Myrella L Cruz, Mayra I Gonzalez Torres, Gladys Chompre, Angel A Isidro, Caroline B Appleyard
ABSTRAKT
The probiotic mixture VSL#3 attenuates colitis in patients with Inflammatory Bowel Disease (IBD) and in animal models of this condition, but the mechanisms involved are incompletely understood. VSL#3 alters macrophage morphology and secretory profile in vitro in a polarization-dependent manner. We examined the effect of VSL#3 on macrophages in acute trinitrobenzene sulfonic acid-induced colitis. Rats were randomized to normal, colitis, or colitis+VSL#3 groups. After sacrifice, the colons were evaluated for macroscopic and microscopic damage. Serum cytokine levels were measured, and microbiome analysis undertaken. Total and M1 colonic macrophages, and total and proliferating hepatic macrophages were assessed by double immunofluorescence staining. Colitis+VSL#3 rats had lower macroscopic damage, with less microscopic damage in the proximal colon, compared with colitis alone. Colitis significantly increased colonic macrophage infiltration, which was significantly reduced by VSL#3 treatment. VSL#3 did not decrease the colitis-induced surge of colonic M1 macrophages or hepatic macrophages. VSL#3 reduced colitis-induced serum cytokine levels, and induced restoration of colonic transcript levels for pro-inflammatory, anti-inflammatory, and barrier proteins to, or past, normal levels. Fecal bacteria distribution changed between groups. In summary, the probiotic VSL#3 reduces colitis severity, colonic macrophage infiltration, and serum cytokine levels, but does not dampen the pro-inflammatory phenotype of M1 macrophages.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
MILLIPLEX® Rat Cytokine/Chemokine Panel, Configurable Rat Cytokine/Chemokine 27-Plex Panel
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %