Merck
Stimulus duration and response time independently influence the kinetics of lytic cycle reactivation of Epstein-Barr virus.
Journal of virology (2009-08-07)
Jill Countryman, Lyndle Gradoville, Sumita Bhaduri-McIntosh, Jianjiang Ye, Lee Heston, Sarah Himmelfarb, Duane Shedd, George Miller
ABSTRAKT
Epstein-Barr virus (EBV) can be reactivated from latency into the lytic cycle by many stimuli believed to operate by different mechanisms. Cell lines containing EBV differ in their responses to inducing stimuli, yet all stimuli require de novo protein synthesis (44). A crucial step preliminary to identifying these proteins and determining when they are required is to measure the duration of stimulus and response time needed for activation of expression of EBV BRLF1 and BZLF1, the earliest viral indicators of reactivation. Here we show, with four EBV-containing cell lines that respond to different inducing agents, that stimuli that are effective at reactivating EBV can be divided into two main groups. The histone deacetylase inhibitors sodium butyrate and trichostatin A require a relatively long period of exposure, from 2 to 4 h or longer. Phorbol esters, anti-immunoglobulin G (anti-IgG), and, surprisingly, 5-aza-2'-deoxycytidine require short exposures of 15 min or less. The cell/virus background influences the response time. Expression of the EBV BZLF1 and BRLF1 genes can be detected before 2 h in Akata cells treated with anti-IgG, but both long- and short-duration stimuli required 4 or more hr to activate BZLF1 and BRLF1 expression in HH514-16, Raji, or B95-8 cells. Thus, stimulus duration and response time are independent variables. Neither stimulus duration nor response time can be predicted by the number of cells activated into the lytic cycle. These experiments shed new light on the earliest events leading to lytic cycle reactivation of EBV.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Phorbol-12-myristate-13-acetate, Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.