Merck
Loss of CSMD1 expression disrupts mammary duct formation while enhancing proliferation, migration and invasion.
Oncology reports (2017-05-24)
Mohamed Kamal, Deborah L Holliday, Ewan E Morrison, Valerie Speirs, Carmel Toomes, Sandra M Bell
ABSTRAKT
The CUB and sushi multiple domains 1 (CSMD1) gene maps to chromosome 8p23, a region deleted in many cancers. Loss of CSMD1 expression is associated with poor prognosis in breast cancer suggesting that it acts as a tumour suppressor in this cancer. However, the function of CSMD1 is largely unknown. Herein, we investigated CSMD1 functions in cell line models. CSMD1 expression was suppressed in MCF10A and LNCaP cells using short hairpin RNA. Functional assays were performed focusing on the 'normal' MCF10A cell line. Suppression of CSMD1 significantly increased the proliferation, cell migration and invasiveness of MCF10A cells compared to shcontrols. shCSMD1 cells also showed significantly reduced adhesion to Matrigel and fibronectin. In a three-dimensional Matrigel model of MCF10A cells, reduced CSMD1 expression resulted in the development of larger and more poorly differentiated breast acini-like structures that displayed impaired lumen formation. Loss of CSMD1 expression disrupts a model of mammary duct formation while enhancing proliferation, migration and invasion. Our data suggest that CSMD1 is involved in the suppression of a transformed phenotype.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human CSMD1