Merck
Investigation of Thermally Induced Degradation in CH
Scientific reports (2017-07-07)
Nam-Koo Kim, Young Hwan Min, Seokhwan Noh, Eunkyung Cho, Gitaeg Jeong, Minho Joo, Seh-Won Ahn, Jeong Soo Lee, Seongtak Kim, Kyuwook Ihm, Hyungju Ahn, Yoonmook Kang, Hae-Seok Lee, Donghwan Kim
ABSTRAKT
In this study, we employ a combination of various in-situ surface analysis techniques to investigate the thermally induced degradation processes in MAPbI
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-tert-Butylpyridine, 98%