- Potential application of SERS for arsenic speciation in biological matrices.
Potential application of SERS for arsenic speciation in biological matrices.
Analytical and bioanalytical chemistry (2017-07-02)
Mingwei Yang, Shannon Matulis, Lawrence H Boise, Anthony J McGoron, Yong Cai
PMID28667385
ABSTRAKT
Speciation of arsenic is usually carried out using chromatography-based methods coupled with spectroscopic determination; however, the inevitable procedures involving sample preparation and separation could potentially alter the integrity of the arsenic metabolites present in biological samples. Surface-enhanced Raman spectroscopy (SERS) could be a promising alternative for providing a reliable arsenic analysis under the influence of a cellular matrix. A method for arsenic speciation using SERS in cellular matrix was developed in this study and four arsenicals were selected, including arsenite (As