Merck
Anti-Inflammatory and Analgesic Activities of the Complex of Flavonoids from Lychnis chalcedonica L.
Bulletin of experimental biology and medicine (2017-07-21)
Yu V Nesterova, T N Povet'eva, L N Zibareva, N I Suslov, E P Zueva, S G Aksinenko, O G Afanas'eva, S G Krylova, E N Amosova, O Yu Rybalkina, K A Lopatina
ABSTRAKT
Anti-inflammatory and analgesic activities of the complex of flavonoids from Lychnis chalcedonica L. were studied in the models of acute aseptic inflammation induced by carrageenan, histamine, and serotonin and acetic acid-induced painful chemical stimulation. It is demonstrated that course treatment with flavonoids derived from Lychnis chalcedonica L. produced a stable pharmacological effect comparable with that of the reference anti-inflammatory drug diclofenac.