Merck
Clinical aspects of urea transporters.
Sub-cellular biochemistry (2014-10-10)
Jianhua Ran, Hongkai Wang, Tinghai Hu
ABSTRAKT
Jk antigens, which were identified as urea transporter B (UT-B) in the plasma membrane of erythrocytes, and which determine the Kidd blood type in humans, are involved in transfusion medicine, and even in organ transplantation. The Jk(a-b-) blood type is a consequence of a silent Slc14A1 gene caused by various mutations related to lineage. In addition, the specific mutations related to hypertension and metabolic syndrome cannot be ignored. Genome-wide association studies established Slc14A1 as a related gene of bladder cancer and some genotypes are associated with higher morbidity. This chapter aims to introduce the clinical significance of urea transporters.