Merck
  • Mixed-substrate (glycerol tributyrate and fibrin) zymography for simultaneous detection of lipolytic and proteolytic enzymes on a single gel.
Mixed-substrate (glycerol tributyrate and fibrin) zymography for simultaneous detection of lipolytic and proteolytic enzymes on a single gel.
Electrophoresis (2009-06-23)
Nack-Shick Choi, Jong Hyun Choi, Bo-Hye Kim, Yun-Jon Han, Joong Su Kim, Seung-Goo Lee, Jae Jun Song
ABSTRAKT
A new zymography method for simultaneous detection of two different enzymatic activities (lipolytic and proteolytic) using a single SDS-containing or native-conformation gel and a mixed-substrate (glycerol tributyrate and fibrin) (MS)(1) gel was developed. After routine electrophoresis, SDS in the gel was removed by treatment with Triton X-100. Gel proteins were electrotransferred to the MS gel. To visualize lipolytic activity, the MS gel was incubated at 37 degrees C (for 6 or 24 h) until clear bands against an opaque background were observed. To detect proteolytic activity, the same MS gel was stained with Coomassie brilliant blue. Using this method, we show that six lipolytic enzymes from Staphylococcus pasteuri NJ-1 and four proteolytic enzymes from two Bacillus strains, B. licheniformis DJ-2 and B. licheniformis NJ-5, isolated from soil, can be simultaneously detected.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid