Merck
Sonochemical synthesis, formation mechanism, and solar cell application of tellurium nanoparticles.
Ultrasonics sonochemistry (2017-07-25)
Mehdi Mousavi-Kamazani, Reza Rahmatolahzadeh, Seyed Amin Shobeiri, Farshad Beshkar
ABSTRAKT
Tellurium inherently tends to form 1-D structures and while the 0-D Te nanostructures have better properties and applications in solar cell. In the present study, 0-D Te nanostructures including rice-like and spherical nanoparticles with the particle size of 15-40nm were successfully synthesized via a facile sonochemical method. In the absence of ultrasonic irradiation nanorods were produced while performing the reaction under ultrasonic waves (at 200W for 30min) led to the formation of nanoparticles. Finally, the efficiency of various as-synthesized Te nanostructures in quantum dot-sensitized solar cells (QDSSCs) were evaluated. Using rice-like nanoparticles led to increase in J
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Valeronitrile, 99.5%
Sigma-Aldrich
4-tert-Butylpyridine, 98%