Merck
Extensive Metabolic Remodeling Differentiates Non-pathogenic and Pathogenic Growth Forms of the Dimorphic Pathogen
Frontiers in cellular and infection microbiology (2017-09-02)
Shivani Pasricha, James I MacRae, Hwa H Chua, Jenny Chambers, Kylie J Boyce, Malcolm J McConville, Alex Andrianopoulos
ABSTRAKT
Fungal infections are an increasing public health problem, particularly in immunocompromised individuals. While these pathogenic fungi show polyphyletic origins with closely related non-pathogenic species, many undergo morphological transitions to produce pathogenic cell types that are associated with increased virulence. However, the characteristics of these pathogenic cells that contribute to virulence are poorly defined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trifluoroacetamide, 97%