Merck
Preclinical studies for a phase 1 clinical trial of autologous hematopoietic stem cell gene therapy for sickle cell disease.
Cytotherapy (2017-07-25)
Fabrizia Urbinati, Jennifer Wherley, Sabine Geiger, Beatriz Campo Fernandez, Michael L Kaufman, Aaron Cooper, Zulema Romero, Filippo Marchioni, Lilith Reeves, Elizabeth Read, Barbara Nowicki, Elke Grassman, Shivkumar Viswanathan, Xiaoyan Wang, Roger P Hollis, Donald B Kohn
ABSTRAKT
Gene therapy by autologous hematopoietic stem cell transplantation (HSCT) represents a new approach to treat sickle cell disease (SCD). Optimization of the manufacture, characterization and testing of the transduced hematopoietic stem cell final cell product (FCP), as well as an in depth in vivo toxicology study, are critical for advancing this approach to clinical trials. Data are shown to evaluate and establish the feasibility of isolating, transducing with the Lenti/β Primary and secondary transplantation did not reveal any toxicity from the lentiviral vector. Additionally, vector integration site analysis of murine and human BM cells did not show any clonal skewing caused by insertion of the Lenti/β We present here a complete protocol, thoroughly optimized to manufacture, characterize and establish safety of a FCP for gene therapy of SCD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)