Merck
Cytokine profile of NALT during acute stress and its possible effect on IgA secretion.
Immunology letters (2017-05-05)
Juan Manuel Gutiérrez-Meza, Rosa Adriana Jarillo-Luna, Victor Rivera-Aguilar, Angel Miliar-García, Rafael Campos-Rodríguez
ABSTRAKT
Stress stimuli affect the immune system responses that occur at mucosal membranes, particularly IgA secretion. It has been suggested that acute stress increases the levels of IgA and that sympathetic innervation plays an important role in this process. We herein explore in a murine model how acute stress affects the Th1/Th2/Treg cytokine balance in NALT, and the possible role of glucocorticoids in this effect. Nine-week-old male CD1 mice were divided into three groups: unstressed (control), stressed (subjected to 4h of immobilization), and stressed after pretreatment with a single dose of the corticosterone receptor antagonist RU-486. The parameters evaluated included plasma corticosterone and epinephrine, IgA levels in nasal fluid (by ELISA), the percentage of CD19
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IgA, Kappa from murine myeloma, clone TEPC 15, purified immunoglobulin, buffered aqueous solution