Merck
Design, Synthesis and Biological Evaluation of novel Hedgehog Inhibitors for treating Pancreatic Cancer.
Scientific reports (2017-05-12)
Vinod Kumar, Amit Kumar Chaudhary, Yuxiang Dong, Haizhen A Zhong, Goutam Mondal, Feng Lin, Virender Kumar, Ram I Mahato
ABSTRAKT
Hedgehog (Hh) pathway is involved in epithelial-mesenchymal transition (EMT) and cancer stem cell (CSC) maintenance resulting in tumor progression. GDC-0449, an inhibitor of Hh pathway component smoothened (Smo) has shown promise in the treatment of various cancers including pancreatic cancer. However, the emergence of resistance during GDC-0449 treatment with numerous side effects limits its use. Therefore, here we report the design, synthesis and evaluation of novel GDC-0449 analogs using N-[3-(2-pyridinyl) phenyl] benzamide scaffold. Cell-based screening followed by molecular simulation revealed 2-chloro-N
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Di-(2-picolyl)amine, 97%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)