Merck
Dual-Mode Optical Sensing of Histamine at Nanomolar Concentrations in Complex Biological Fluids and Living Cells.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-06-16)
Nilanjan Dey, Asfa Ali, Santosh Podder, Shamik Majumdar, Dipankar Nandi, Santanu Bhattacharya
ABSTRAKT
An easily synthesized fluorescein-based luminescent dye has been utilized for the dual-mode detection of histamine at nanomolar concentrations at pH 7.0 in water. The specific response to histamine was achieved by imidazole-catalyzed 'imine formation' reaction. The protocol was subsequently applied for the estimation of histamine in complex biological milieu such as human blood serum and urine samples. Furthermore, the dose-dependent cellular uptake of histamine and de novo synthesis (by thapsigargin treatment) was visualized in RAW 264.7, a mouse macrophage cell line. We have also developed portable paper strips for rapid, on-site detection of histamine without involving costly instruments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film