Merck
Raman active components of skin cancer.
Biomedical optics express (2017-07-01)
Xu Feng, Austin J Moy, Hieu T M Nguyen, Jason Zhang, Matthew C Fox, Katherine R Sebastian, Jason S Reichenberg, Mia K Markey, James W Tunnell
ABSTRAKT
Raman spectroscopy (RS) has shown great potential in noninvasive cancer screening. Statistically based algorithms, such as principal component analysis, are commonly employed to provide tissue classification; however, they are difficult to relate to the chemical and morphological basis of the spectroscopic features and underlying disease. As a result, we propose the first Raman biophysical model applied to
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)