Merck
  • Kinetic Study of the Aroxyl-Radical-Scavenging Activity of Five Fatty Acid Esters and Six Carotenoids in Toluene Solution: Structure-Activity Relationship for the Hydrogen Abstraction Reaction.
Kinetic Study of the Aroxyl-Radical-Scavenging Activity of Five Fatty Acid Esters and Six Carotenoids in Toluene Solution: Structure-Activity Relationship for the Hydrogen Abstraction Reaction.
The journal of physical chemistry. B (2017-07-25)
Kazuo Mukai, Maya Yoshimoto, Masaharu Ishikura, Shin-Ichi Nagaoka
ABSTRAKT
A kinetic study of the reaction between an aroxyl radical (ArO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl linolenate, ≥98%
Sigma-Aldrich
Ethyl arachidonate, ≥98.5% (GC), liquid