Merck
Hydroethanolic extract of
Iranian journal of basic medical sciences (2017-11-01)
Saeid Abbasi-Maleki, Zahra Mousavi
ABSTRAKT
Studies indicate that major deficiency in the levels of monoaminergic transmitters is a reason for severe depression. On the other hand, it is shown that The mice were intraperitoneally (IP) treated with CT extract (100-400 mg/kg) 1 hr before the TST. To investigate the involvement of monoaminergic systems in antidepressant-like effect, the mice were treated with receptor antagonists 15 min before CT extract treatment (400 mg/kg, IP) and 1 hr before the TST. Findings showed that CT extract (100-400 mg/kg, IP), dose-dependently induced antidepressant-like effect ( Our findings firstly showed that components (especially N-Hexadecanoic acid) of CT extract induce antidepressant-like effects by interaction with dopaminergic (D1 and D2) and serotonergic (5HT1A, 5-HT2A receptors) systems. These findings validate the folk use of CT extract for the management of depression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Chloro-L-phenylalanine