- Role of the phagosomal redox-sensitive TRP channel TRPM2 in regulating bactericidal activity of macrophages.
Role of the phagosomal redox-sensitive TRP channel TRPM2 in regulating bactericidal activity of macrophages.
Journal of cell science (2017-01-14)
Anke Di, Tomohiro Kiya, Haixia Gong, Xiaopei Gao, Asrar B Malik
PMID28082421
ABSTRAKT
Acidification of macrophage phagosomes serves an important bactericidal function. We show here that the redox-sensitive transient receptor potential (TRP) cation channel TRPM2 is expressed in the phagosomal membrane and regulates macrophage bactericidal activity through the activation of phagosomal acidification. Measurement of the TRPM2 current in phagosomes identified TRPM2 as a functional redox-sensitive cation channel localized in the phagosomal membrane. Simultaneous measurements of phagosomal Ca
MATERIAŁY