Merck
  • Specific Combination of Salvianolic Acids As Core Active Ingredients of Danhong Injection for Treatment of Arterial Thrombosis and Its Derived Dry Gangrene.
Specific Combination of Salvianolic Acids As Core Active Ingredients of Danhong Injection for Treatment of Arterial Thrombosis and Its Derived Dry Gangrene.
Frontiers in pharmacology (2017-07-01)
Tiechan Zhao, Lianying Chang, Boyong Zhang, Ming Lu, Xiaoyi Wang, John O Orgah, Yuefei Wang, Xiaoxuan Tian, Jing Yang, Guanwei Fan, Boli Zhang, Yan Zhu
ABSTRAKT
Although single-targeting anti-platelet agents are used extensively in clinics, their limitations in resistance and bleeding have started a trend of combination therapy. Danhong injection (DHI) is a widely prescribed injection medicine for cardiovascular and cerebrovascular diseases in China. However, its precise clinical efficacy and functional components remain unexplored. In this study, we investigated the anti-thrombotic role and its chemical basis of DHI. In a photochemically-induced thrombosis model, DHI effectively dissolved thrombus and ameliorated its derived dry gangrene. DHI inhibited multiple GPCR agonists-induced platelet adhesion, aggregation and downstream Ca
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%