Merck
Allosteric Control of Substrate Specificity of the
Frontiers in chemistry (2017-07-05)
Ana C Ebrecht, Ligin Solamen, Benjamin L Hill, Alberto A Iglesias, Kenneth W Olsen, Miguel A Ballicora
ABSTRAKT
The substrate specificity of enzymes is crucial to control the fate of metabolites to different pathways. However, there is growing evidence that many enzymes can catalyze alternative reactions. This promiscuous behavior has important implications in protein evolution and the acquisition of new functions. The question is how the undesirable outcomes of
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine-5′-diphosphoglucose disodium salt, ≥93%
Sigma-Aldrich
Uridine 5′-diphosphoglucose disodium salt, ≥98.0% (HPLC)