Merck
bisFabs: Tools for rapidly screening hybridoma IgGs for their activities as bispecific antibodies.
mAbs (2017-01-27)
Sanket Patke, Ji Li, Peiyin Wang, Dion Slaga, Jennifer Johnston, Sunil Bhakta, Siler Panowski, Liping L Sun, Teemu Junttila, Justin M Scheer, Diego A Ellerman
ABSTRAKT
Bispecific antibodies are a growing class of therapeutic molecules. Many of the current bispecific formats require DNA engineering to convert the parental monoclonal antibodies into the final bispecific molecules. We describe here a method to generate bispecific molecules from hybridoma IgGs in 3-4 d using chemical conjugation of antigen-binding fragments (Fabs) (bisFabs). Proteolytic digestion conditions for each IgG isotype were analyzed to optimize the yield and quality of the final conjugates. The resulting bisFabs showed no significant amounts of homodimers or aggregates. The predictive value of murine bisFabs was tested by comparing the T-cell redirected cytotoxic activity of a panel of antibodies in either the bisFab or full-length IgG formats. A variety of antigens with different structures and expression levels was used to extend the comparison to a wide range of binding geometries and antigen densities. The activity observed for different murine bisFabs correlated with those observed for the full-length IgG format across multiple different antigen targets, supporting the use of bisFabs as a screening tool. Our method may also be used for the screening of bispecific antibodies with other mechanisms of action, allowing for a more rapid selection of lead therapeutic candidates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1′-(Methylenedi-4,1-phenylene)bismaleimide, 95%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein