Merck
Preparation of clinical-scale
Journal of labelled compounds & radiopharmaceuticals (2017-02-10)
Mohini Guleria, Tapas Das, Chandan Kumar, Jeyachitra Amirdhanayagam, Haladhar D Sarma, Sharmila Banerjee
ABSTRAKT
Rituximab is a monoclonal chimeric antibody, which has been approved by the US Food and Drug Administration for immunotherapy of non-Hodgkin lymphoma. Bexxar and Zevalin are the two other approved radiolabeled antibodies for radioimmunotherapy of non-Hodgkin lymphoma; however, they are of murine origin that reduces their treatment efficacy. So as to circumvent this, efforts have been made to radiolabel Rituximab with various therapeutic radioisotopes. In the present study, an effort has been made to optimize the conjugation (bifunctional chelating agent and antibody) and radiolabeling procedures for the preparation of clinical-scale
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Arsenazo III, calcium-sensitive dye