- One-pot synthesis of Au@TiO
One-pot synthesis of Au@TiO
Journal of colloid and interface science (2017-07-04)
Hang Sun, Qinrong He, Ping She, Shan Zeng, Kongliang Xu, Jiayi Li, Song Liang, Zhenning Liu
PMID28672267
ABSTRAKT
Natural biological systems often use hollow structures to decrease reflection and achieve high solar light utilization. Herein, bio-inspired Au@TiO
MATERIAŁY