Merck
Nanosized CaP-silk fibroin-PCL-PEG-PCL/PCL based bilayer membranes for guided bone regeneration.
Materials science & engineering. C, Materials for biological applications (2017-09-04)
Sibel Türkkan, A Engin Pazarçeviren, Dilek Keskin, Nesrin E Machin, Özgür Duygulu, Ayşen Tezcaner
ABSTRAKT
Guided bone regeneration (GBR) concept has been developed to prevent the formation of non-functional scar tissue layer on defect site by undertaking barrier role. In this study, a new bilayer membrane which consisted of one layer of electrospun silk fibroin/PCL-PEG-PCL incorporating nanocalcium phosphate (SPCA)