Merck
  • Simultaneous quantitation of endogenous estrone, 17β-estradiol, and estriol in human serum by isotope-dilution liquid chromatography-tandem mass spectrometry for clinical laboratory applications.
Simultaneous quantitation of endogenous estrone, 17β-estradiol, and estriol in human serum by isotope-dilution liquid chromatography-tandem mass spectrometry for clinical laboratory applications.
Analytical and bioanalytical chemistry (2017-02-23)
Qiaoxuan Zhang, Liqiao Han, Jianbing Wang, Haibiao Lin, Peifeng Ke, Junhua Zhuang, Xianzhang Huang
ABSTRAKT
Estrogen measurements are important in the assessment of female reproductive function and have expanding roles in other fields. A simple, accurate, highly sensitive and specific isotope-dilution liquid chromatography-tandem mass spectrometry method was developed and evaluated to simultaneously measure three endogenous estrogens in serum: estrone (E
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Hyodeoxycholic acid, ≥98%