Merck
Synthesis of Copper Oxide/Graphite Composite for High-Performance Rechargeable Battery Anode.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-06-28)
Sanghun Cho, Yong-Keon Ahn, Zhenxing Yin, Duck-Jae You, Hyunjin Kim, Yuanzhe Piao, Jeeyoung Yoo, Youn Sang Kim
ABSTRAKT
A novel copper oxide/graphite composite (GCuO) anode with high capacity and long cycle stability is proposed. A simple, one-step synthesis method is used to prepare the GCuO, through heat treatment of the Cu ion complex and pristine graphite. The gases generated during thermal decomposition of the Cu ion complex (H
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Octylamine, 99%