- Pilot application of lipoxin A
Pilot application of lipoxin A
Experimental and therapeutic medicine (2017-10-01)
Xia Kong, Sheng-Hua Wu, Li Zhang, Xiao-Qing Chen
PMID28962156
ABSTRAKT
Previous studies have demonstrated that lipoxin A
Previous studies have demonstrated that lipoxin A