- Template-Assisted Preparation of Micrometric Suspended Membrane Lattices of Photoluminescent and Non-Photoluminescent Polymers by Capillarity-Driven Solvent Evaporation: Application to Microtagging.
Template-Assisted Preparation of Micrometric Suspended Membrane Lattices of Photoluminescent and Non-Photoluminescent Polymers by Capillarity-Driven Solvent Evaporation: Application to Microtagging.
Scientific reports (2017-08-23)
Giovanni Polito, Valentina Robbiano, Chiara Cozzi, Franco Cacialli, Giuseppe Barillaro
PMID28827776
ABSTRAKT
In this work, the bottom-up template-assisted preparation of high-density lattices (up to 11 · 10