Merck
Soluble PD-L1 as a Biomarker in Malignant Melanoma Treated with Checkpoint Blockade.
Cancer immunology research (2017-05-20)
Jun Zhou, Kathleen M Mahoney, Anita Giobbie-Hurder, Fengmin Zhao, Sandra Lee, Xiaoyun Liao, Scott Rodig, Jingjing Li, Xinqi Wu, Lisa H Butterfield, Matthias Piesche, Michael P Manos, Lauren M Eastman, Glenn Dranoff, Gordon J Freeman, F Stephen Hodi
ABSTRAKT
Blockade of the pathway including programmed death-ligand 1 (PD-L1) and its receptor programmed cell death protein 1 (PD-1) has produced clinical benefits in patients with a variety of cancers. Elevated levels of soluble PD-L1 (sPD-L1) have been associated with worse prognosis in renal cell carcinoma and multiple myeloma. However, the regulatory roles and function of sPD-L1 particularly in connection with immune checkpoint blockade treatment are not fully understood. We identified four splice variants of PD-L1 in melanoma cells, and all of them are secreted. Secretion of sPD-L1 resulted from alternate splicing activities, cytokine induction, cell stress, cell injury, and cell death in melanoma cells. Pretreatment levels of sPD-L1 were elevated in stage IV melanoma patient sera compared with healthy donors. High pretreatment levels of sPD-L1 were associated with increased likelihood of progressive disease in patients treated by CTLA-4 or PD-1 blockade. Although changes in circulating sPD-L1 early after treatment could not distinguish responders from those with progressive disease, after five months of treatment by CTLA-4 or PD-1 blockade patients who had increased circulating sPD-L1 had greater likelihood of developing a partial response. Induction of sPD-L1 was associated with increased circulating cytokines after CTLA-4 blockade but not following PD-1 blockade. Circulating sPD-L1 is a prognostic biomarker that may predict outcomes for subgroups of patients receiving checkpoint inhibitors.