Merck
Investigation of protein synthesis in
Biology open (2017-06-24)
Lisa P Deliu, Abhishek Ghosh, Savraj S Grewal
ABSTRAKT
Translational control of gene expression is an important regulator of growth, homeostasis and aging in
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
cOmplete™ Mini koktajl inhibitorów proteaz, Tablets provided in EASYpacks