Merck
TUSC3 induces autophagy in human non-small cell lung cancer cells through Wnt/β-catenin signaling.
Oncotarget (2017-09-09)
Yun Peng, Jun Cao, Xiao-Yi Yao, Jian-Xin Wang, Mei-Zuo Zhong, Ping-Ping Gan, Jian-Huang Li
ABSTRAKT
We investigated the effects of tumor suppressor candidate 3 (
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TUSC3