- The pro-inflammatory signalling regulator Stat4 promotes vasculogenesis of great vessels derived from endothelial precursors.
The pro-inflammatory signalling regulator Stat4 promotes vasculogenesis of great vessels derived from endothelial precursors.
Nature communications (2017-03-04)
Zhao-Zheng Meng, Wei Liu, Yu Xia, Hui-Min Yin, Chi-Yuan Zhang, Dan Su, Li-Feng Yan, Ai-Hua Gu, Yong Zhou
PMID28256502
ABSTRAKT
Vasculogenic defects of great vessels (GVs) are a major cause of congenital cardiovascular diseases. However, genetic regulators of endothelial precursors in GV vasculogenesis remain largely unknown. Here we show that Stat4, a transcription factor known for its regulatory role of pro-inflammatory signalling, promotes GV vasculogenesis in zebrafish. We find stat4 transcripts highly enriched in nkx2.5
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-STAT1α antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution